Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68211208-68211451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLN6
Entrez Gene ID
54982
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Gene Aliases
CLN4A, HsT18960, nclf
UniGene
Hs.584921
Gene Chromosome Location
Chr.15: 68206992-68229742 on Build GRCh38
Transcripts
NM_017882.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1374250 c.535T>C p.C179R Chr.15: 68211270-68211270 on Build GRCh38
3794351 c.489C>T p.I163I Chr.15: 68211316-68211316 on Build GRCh38
376438 c.532C>G p.H178D Chr.15: 68211273-68211273 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs763901402 Chr.15: 68211271-68211271 on Build GRCh38
rs761574362 Chr.15: 68211352-68211352 on Build GRCh38
rs912430944 Chr.15: 68211390-68211390 on Build GRCh38
rs547002745 Chr.15: 68211289-68211289 on Build GRCh38
rs774500792 Chr.15: 68211233-68211233 on Build GRCh38
rs760584097 Chr.15: 68211252-68211252 on Build GRCh38
rs116956347 Chr.15: 68211357-68211357 on Build GRCh38
rs762902907 Chr.15: 68211306-68211307 on Build GRCh38
rs563851812 Chr.15: 68211413-68211413 on Build GRCh38
rs121908079 Chr.15: 68211292-68211294 on Build GRCh38
rs786205066 Chr.15: 68211258-68211258 on Build GRCh38
rs200775021 Chr.15: 68211333-68211333 on Build GRCh38
rs369649575 Chr.15: 68211346-68211346 on Build GRCh38
rs772888484 Chr.15: 68211403-68211403 on Build GRCh38
rs776617600 Chr.15: 68211243-68211243 on Build GRCh38
rs757916285 Chr.15: 68211310-68211310 on Build GRCh38
rs756942559 Chr.15: 68211286-68211286 on Build GRCh38
rs200979467 Chr.15: 68211347-68211347 on Build GRCh38
rs374426304 Chr.15: 68211359-68211359 on Build GRCh38
rs1011355697 Chr.15: 68211407-68211407 on Build GRCh38
rs779685727 Chr.15: 68211315-68211315 on Build GRCh38
rs548821903 Chr.15: 68211230-68211230 on Build GRCh38
rs767578507 Chr.15: 68211247-68211247 on Build GRCh38
rs555379379 Chr.15: 68211327-68211327 on Build GRCh38
rs747926816 Chr.15: 68211331-68211331 on Build GRCh38
rs763264386 Chr.15: 68211228-68211228 on Build GRCh38
rs778284215 Chr.15: 68211328-68211328 on Build GRCh38
rs371794062 Chr.15: 68211332-68211332 on Build GRCh38
rs200117997 Chr.15: 68211356-68211356 on Build GRCh38
rs571940397 Chr.15: 68211330-68211330 on Build GRCh38
rs748682605 Chr.15: 68211316-68211316 on Build GRCh38
rs750081097 Chr.15: 68211296-68211296 on Build GRCh38
rs991076289 Chr.15: 68211256-68211256 on Build GRCh38
rs529424184 Chr.15: 68211414-68211414 on Build GRCh38
rs916824303 Chr.15: 68211284-68211286 on Build GRCh38
rs759592282 Chr.15: 68211237-68211237 on Build GRCh38
rs773659823 Chr.15: 68211226-68211226 on Build GRCh38
rs763954715 Chr.15: 68211349-68211349 on Build GRCh38
rs202046680 Chr.15: 68211326-68211326 on Build GRCh38
rs772189210 Chr.15: 68211338-68211338 on Build GRCh38
rs941060139 Chr.15: 68211293-68211293 on Build GRCh38
rs776501944 Chr.15: 68211351-68211351 on Build GRCh38
rs752672375 Chr.15: 68211250-68211250 on Build GRCh38
rs1039634789 Chr.15: 68211378-68211378 on Build GRCh38
rs919844747 Chr.15: 68211431-68211431 on Build GRCh38
rs1025121989 Chr.15: 68211251-68211251 on Build GRCh38
rs753538013 Chr.15: 68211283-68211283 on Build GRCh38

View Full Product Details