Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR45
Entrez Gene ID
11152
Gene Name
WD repeat domain 45
Gene Aliases
JM5, NBIA4, NBIA5, WDRX1, WIPI-4, WIPI4
Chromosome Location
Chr.X: 49074433 - 49101121 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029896.1 NP_001025067.1 6 - 7 615 78
NM_007075.3 NP_009006.2 7 - 8 885 78
GenBank mRNA AJ005897.1 - 6 - 7 571 78
AK023344.1 - 4 - 5 1593 78
AK225122.1 - 6 - 7 594 78
AK315717.1 - 6 - 7 602 78
AY691428.1 - 5 - 6 443 78
BC000464.2 - 6 - 7 609 78
BC003037.1 - 6 - 7 609 78
BC069206.1 - 7 - 8 885 78
BQ068083.1 - 6 - 7 615 78
CR456725.1 - 5 - 6 443 78

View Full Product Details