The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289160.1 | NP_001276089.1 | 5 - 6 | 1066 | 138 |
NM_001289161.1 | NP_001276090.1 | 5 - 6 | 1036 | 138 | |
NM_005931.4 | NP_005922.2 | 5 - 6 | 1165 | 138 | |
GenBank mRNA | AF021221.1 | - | 4 - 5 | 951 | 138 |
AF021222.1 | - | 4 - 5 | 951 | 138 | |
AF021223.1 | - | 4 - 5 | 951 | 138 | |
AF021224.1 | - | 4 - 5 | 951 | 138 | |
AF021225.1 | - | 4 - 5 | 951 | 138 | |
AF021226.1 | - | 4 - 5 | 951 | 138 | |
AK291927.1 | - | 5 - 6 | 1163 | 138 | |
AK300791.1 | - | 5 - 6 | 1066 | 138 | |
AK303655.1 | - | 4 - 5 | 1300 | 138 | |
AK314228.1 | - | 5 - 6 | 1070 | 138 | |
AY885251.1 | - | 6 - 7 | 1117 | 138 | |
BC044218.1 | - | 5 - 6 | 1000 | 138 | |
X91625.1 | - | 5 - 6 | 1026 | 138 |