Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMRTC1
Entrez Gene ID
63947
Gene Name
DMRT like family C1
Gene Aliases
-
Chromosome Location
Chr.X: 72872025 - 72943653 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033053.2 NP_149042.2 5 - 6 558 162
XM_005262287.3 XP_005262344.1 6 - 7 637 162
XM_005262288.3 XP_005262345.1 6 - 7 662 162
XM_017029725.1 XP_016885214.1 6 - 7 668 162
GenBank mRNA AV753283.1 - - 103 162
AW205069.1 - 1 - 2 118 162
BC029799.1 - 5 - 6 982 162
BC047596.1 - 4 - 5 506 162

Gene Symbol
DMRTC1B
Entrez Gene ID
728656
Gene Name
DMRT like family C1B
Gene Aliases
-
Chromosome Location
Chr.X: 72777170 - 72848802 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080851.1 NP_001074320.1 5 - 6 558 162
XM_006724684.2 XP_006724747.1 6 - 7 637 162
XM_006724685.3 XP_006724748.1 6 - 7 662 162
XM_017029776.1 XP_016885265.1 6 - 7 668 162

View Full Product Details