The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153705.4 | NP_714916.3 | 6 - 7 | 1361 | 125 |
XM_011542621.2 | XP_011540923.1 | 6 - 7 | 1334 | 125 | |
XM_011542623.2 | XP_011540925.1 | 4 - 5 | 914 | 125 | |
GenBank mRNA | AF533708.1 | - | 6 - 7 | 1320 | 125 |
AK098206.1 | - | 2 - 3 | 326 | 125 | |
AK313534.1 | - | 6 - 7 | 1223 | 125 | |
AY358616.1 | - | 5 - 6 | 1430 | 125 | |
BC036526.2 | - | 6 - 7 | 1315 | 125 |