The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136035.2 | NP_001129507.1 | 2 - 3 | 376 | 97 |
NM_001142554.1 | NP_001136026.1 | 1 - 2 | 497 | 97 | |
NM_017722.3 | NP_060192.1 | 1 - 2 | 497 | 97 | |
XM_011528124.1 | XP_011526426.1 | 2 - 3 | 295 | 97 | |
XM_017026944.1 | XP_016882433.1 | 3 - 4 | 819 | 97 | |
XM_017026945.1 | XP_016882434.1 | 3 - 4 | 819 | 97 | |
XM_017026947.1 | XP_016882436.1 | 3 - 4 | 818 | 97 | |
GenBank mRNA | AF196479.1 | - | 1 - 2 | 247 | 97 |
AK000251.1 | - | 2 - 3 | 335 | 97 | |
AK292377.1 | - | 2 - 3 | 376 | 97 | |
AK295245.1 | - | 2 - 3 | 276 | 97 | |
BC002492.2 | - | 1 - 2 | 327 | 97 | |
BC018302.1 | - | 1 - 2 | 492 | 97 | |
BC040126.1 | - | 1 - 2 | 366 | 97 | |
DC344734.1 | - | 1 - 2 | 497 | 97 |