The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305077.1 | NP_001292006.1 | 4 - 5 | 501 | 154 |
NM_001305078.1 | NP_001292007.1 | 4 - 5 | 500 | 154 | |
NM_001305080.1 | NP_001292009.1 | 4 - 5 | 504 | 139 | |
NM_001305081.1 | NP_001292010.1 | 4 - 5 | 501 | 151 | |
NM_001305082.1 | NP_001292011.1 | 4 - 5 | 649 | 154 | |
NM_001305083.1 | NP_001292012.1 | 4 - 5 | 501 | 139 | |
NM_015654.4 | NP_056469.2 | 4 - 5 | 504 | 154 | |
XM_005257212.4 | XP_005257269.1 | 3 - 4 | 343 | 154 | |
XM_006721813.3 | XP_006721876.1 | 3 - 4 | 791 | 154 | |
XM_017024437.1 | XP_016879926.1 | 4 - 5 | 430 | 154 | |
GenBank mRNA | AK225602.1 | - | 4 - 5 | 374 | 154 |
AK312958.1 | - | 4 - 5 | 374 | 154 | |
AL050269.1 | - | 4 - 5 | 381 | 154 | |
AL537925.3 | - | 4 - 5 | 581 | 154 | |
AY632082.1 | - | 3 - 4 | 340 | 154 | |
BC004195.2 | - | 4 - 5 | 410 | 154 | |
BC004225.2 | - | 4 - 5 | 399 | 154 | |
BM909550.1 | - | 4 - 5 | 403 | 139 | |
BP231776.1 | - | 4 - 5 | 422 | 154 | |
BU159218.1 | - | 4 - 5 | 399 | 151 |