Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC8A3
Entrez Gene ID
6547
Gene Name
solute carrier family 8 member A3
Gene Aliases
NCX3
Chromosome Location
Chr.14: 70044217 - 70189626 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058240.3 NP_489479.1 2 - 3 2539 103
NM_182932.2 NP_891977.1 2 - 3 2539 103
NM_183002.2 NP_892114.1 2 - 3 2539 103
XM_017021606.1 XP_016877095.1 2 - 3 1911 103
XM_017021607.1 XP_016877096.1 2 - 3 2404 103
XM_017021608.1 XP_016877097.1 2 - 3 2165 103
XM_017021609.1 XP_016877098.1 2 - 3 2404 103
GenBank mRNA AF510501.1 - 2 - 3 2539 103
AF510502.1 - 2 - 3 2539 103
AJ304853.1 - 1 - 2 1847 103
AJ508602.1 - 1 - 2 1335 103
BC036783.1 - 2 - 3 2342 103
BC150183.1 - 1 - 2 1862 103
BC150193.1 - 1 - 2 1862 103

View Full Product Details