Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location
Chr.6: 56457987 - 56954671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144769.2 NP_001138241.1 33 - 34 4655 148
NM_001144770.1 NP_001138242.1 31 - 32 4326 148
NM_001723.5 NP_001714.1 20 - 21 3210 148
NM_015548.4 NP_056363.2 20 - 21 3210 148
NM_183380.3 NP_899236.1 30 - 31 4600 148
XM_005249310.3 XP_005249367.1 34 - 35 4749 148
XM_005249315.3 XP_005249372.1 33 - 34 4696 148
XM_005249316.2 XP_005249373.1 26 - 27 4972 148
XM_005249318.2 XP_005249375.1 32 - 33 4410 148
XM_005249319.2 XP_005249376.1 31 - 32 4311 148
XM_005249320.4 XP_005249377.1 31 - 32 4367 148
XM_005249322.4 XP_005249379.1 31 - 32 4262 148
XM_005249323.3 XP_005249380.1 30 - 31 4595 148
XM_005249324.2 XP_005249381.1 20 - 21 3207 148
XM_011514824.2 XP_011513126.1 33 - 34 4660 148
XM_011514825.2 XP_011513127.1 33 - 34 4656 148
XM_011514826.2 XP_011513128.1 30 - 31 4595 148
XM_017011205.1 XP_016866694.1 34 - 35 4759 148
XM_017011206.1 XP_016866695.1 34 - 35 4759 148
XM_017011207.1 XP_016866696.1 34 - 35 4696 148
XM_017011208.1 XP_016866697.1 34 - 35 4759 148
XM_017011209.1 XP_016866698.1 34 - 35 4759 148
XM_017011210.1 XP_016866699.1 34 - 35 4759 148
XM_017011211.1 XP_016866700.1 34 - 35 4759 148
XM_017011212.1 XP_016866701.1 34 - 35 4759 148
XM_017011213.1 XP_016866702.1 34 - 35 4759 148
XM_017011214.1 XP_016866703.1 34 - 35 4759 148
XM_017011215.1 XP_016866704.1 34 - 35 4759 148
XM_017011216.1 XP_016866705.1 34 - 35 4759 148
XM_017011217.1 XP_016866706.1 31 - 32 4340 148
XM_017011219.1 XP_016866708.1 34 - 35 4755 148
XM_017011220.1 XP_016866709.1 33 - 34 4698 148
XM_017011221.1 XP_016866710.1 34 - 35 4755 148
XM_017011222.1 XP_016866711.1 26 - 27 4914 148
XM_017011223.1 XP_016866712.1 34 - 35 4755 148
XM_017011224.1 XP_016866713.1 30 - 31 4595 148
XM_017011225.1 XP_016866714.1 34 - 35 4769 148
GenBank mRNA AF400227.1 - 20 - 21 3102 148
AK299898.1 - 31 - 32 4326 148
BC065536.1 - 20 - 21 3174 148
DA449443.1 - 4 - 5 468 148
L11690.1 - 20 - 21 3204 148
M63618.1 - 10 - 11 1441 148
M69225.1 - 20 - 21 3140 148

View Full Product Details