Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP20-2
Entrez Gene ID
337976
Gene Name
keratin associated protein 20-2
Gene Aliases
KAP20.2
Chromosome Location
Chr.21: 30635264 - 30635461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181616.1 NP_853647.1 1 - 1 58 87
GenBank mRNA AB096957.1 - 1 - 1 86 87
BC111084.1 - 1 - 1 58 87

View Full Product Details