Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMGB3
Entrez Gene ID
3149
Gene Name
high mobility group box 3
Gene Aliases
HMG-2a, HMG-4, HMG2A, HMG4
Chromosome Location
Chr.X: 150980508 - 150990775 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301228.1 NP_001288157.1 5 - 5 1713 169
NM_001301229.1 NP_001288158.1 5 - 5 1604 169
NM_001301231.1 NP_001288160.1 5 - 5 1643 169
NM_005342.3 NP_005333.2 5 - 5 1608 169
GenBank mRNA BX537505.1 - 3 - 3 3002 169
Y10043.1 - 5 - 5 1584 169

View Full Product Details