The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007144.2 | NP_009075.1 | 2 - 3 | 200 | 107 |
XM_005257640.2 | XP_005257697.1 | 3 - 4 | 659 | 107 | |
XM_017025016.1 | XP_016880505.1 | 3 - 4 | 1219 | 107 | |
GenBank mRNA | BC024255.1 | - | 2 - 3 | 196 | 107 |
BX647429.1 | - | 3 - 4 | 659 | 107 |