Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF26
Entrez Gene ID
7574
Gene Name
zinc finger protein 26
Gene Aliases
HEL-179, KOX20
Chromosome Location
Chr.12: 132986348 - 133027102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256279.1 NP_001243208.1 3 - 4 752 132
NM_019591.3 NP_062537.2 2 - 3 653 132
XM_005266182.3 XP_005266239.1 3 - 4 740 132
XM_005266183.3 XP_005266240.1 4 - 5 943 132
XM_011534829.1 XP_011533131.1 3 - 4 856 132
XM_017019920.1 XP_016875409.1 4 - 5 1376 132
XM_017019921.1 XP_016875410.1 4 - 5 839 132
XM_017019922.1 XP_016875411.1 3 - 4 1725 132
XM_017019923.1 XP_016875412.1 3 - 4 1656 132
GenBank mRNA AK122941.1 - 1 - 2 2766 132
AK291243.1 - 2 - 3 620 132
BC046206.2 - 2 - 3 554 132
BU194644.1 - 3 - 4 632 132
DA737586.1 - 2 - 3 599 132
EU668356.1 - 2 - 3 531 132

View Full Product Details