Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 46860763-46861020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYL3
Entrez Gene ID
4634
Gene Name
myosin light chain 3
Gene Aliases
CMH8, MLC-lV/sb, MLC1SB, MLC1V, VLC1, VLCl
UniGene
Hs.517939
Gene Chromosome Location
Chr.3: 46857867-46863483 on Build GRCh38
Transcripts
NM_000258.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6719625 c.220G>A p.G74R Chr.3: 46860763-46860763 on Build GRCh38
3696112 c.186C>T p.D62D Chr.3: 46860797-46860797 on Build GRCh38
5728555 c.157G>A p.E53K Chr.3: 46860960-46860960 on Build GRCh38
4117930 c.217T>C p.Y73H Chr.3: 46860766-46860766 on Build GRCh38
6719629 c.188G>A p.R63H Chr.3: 46860795-46860795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199474702 Chr.3: 46860816-46860816 on Build GRCh38
rs774428107 Chr.3: 46860807-46860807 on Build GRCh38
rs397516277 Chr.3: 46860818-46860818 on Build GRCh38
rs192329378 Chr.3: 46861001-46861001 on Build GRCh38
rs368118534 Chr.3: 46860800-46860800 on Build GRCh38
rs931465740 Chr.3: 46860900-46860900 on Build GRCh38
rs777555567 Chr.3: 46860952-46860952 on Build GRCh38
rs139354105 Chr.3: 46860795-46860795 on Build GRCh38
rs776431941 Chr.3: 46860829-46860829 on Build GRCh38
rs921464842 Chr.3: 46860899-46860899 on Build GRCh38
rs188566911 Chr.3: 46860871-46860871 on Build GRCh38
rs565312070 Chr.3: 46860796-46860796 on Build GRCh38
rs763012715 Chr.3: 46860862-46860862 on Build GRCh38
rs139794067 Chr.3: 46860813-46860813 on Build GRCh38
rs750698483 Chr.3: 46860843-46860843 on Build GRCh38
rs111668273 Chr.3: 46860828-46860828 on Build GRCh38
rs781740041 Chr.3: 46860921-46860921 on Build GRCh38
rs554596780 Chr.3: 46860886-46860886 on Build GRCh38
rs778515428 Chr.3: 46860977-46860977 on Build GRCh38
rs730880960 Chr.3: 46860790-46860790 on Build GRCh38
rs903860936 Chr.3: 46860941-46860941 on Build GRCh38
rs937857201 Chr.3: 46860833-46860833 on Build GRCh38
rs775106506 Chr.3: 46860823-46860823 on Build GRCh38
rs1042816943 Chr.3: 46860934-46860934 on Build GRCh38
rs1039903239 Chr.3: 46860865-46860865 on Build GRCh38
rs749017586 Chr.3: 46860965-46860965 on Build GRCh38
rs1004231349 Chr.3: 46860970-46860970 on Build GRCh38
rs374988162 Chr.3: 46860997-46860997 on Build GRCh38
rs760558240 Chr.3: 46860832-46860832 on Build GRCh38
rs730880953 Chr.3: 46860981-46860981 on Build GRCh38
rs372045422 Chr.3: 46860951-46860951 on Build GRCh38
rs730880955 Chr.3: 46860789-46860789 on Build GRCh38
rs997708651 Chr.3: 46860989-46860989 on Build GRCh38
rs542733581 Chr.3: 46860876-46860876 on Build GRCh38
rs730880954 Chr.3: 46860799-46860799 on Build GRCh38
rs752977796 Chr.3: 46860926-46860926 on Build GRCh38
rs765419653 Chr.3: 46860835-46860835 on Build GRCh38

View Full Product Details