Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46034104-46034361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
UniGene
Hs.648744
Gene Chromosome Location
Chr.17: 46029916-46225374 on Build GRCh38
Transcripts
NM_001193465.1 NM_001193466.1 NM_015443.3 XM_006721823.1 XM_006721824.3 XM_011524628.2 XM_011524629.2 XM_011524630.2 XM_011524631.2 XM_011524632.2 XM_017024488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6990671 c.2569G>C p.E857Q Chr.17: 46034258-46034258 on Build GRCh38
2695617 c.2660C>T p.T887M Chr.17: 46034167-46034167 on Build GRCh38
980240 c.2661G>A p.T887T Chr.17: 46034166-46034166 on Build GRCh38
7168806 c.2626C>T p.R876C Chr.17: 46034201-46034201 on Build GRCh38
980242 c.2572_2573delAG p.S858fs*3 Chr.17: 46034254-46034255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753288133 Chr.17: 46034266-46034266 on Build GRCh38
rs17574604 Chr.17: 46034247-46034247 on Build GRCh38
rs771605434 Chr.17: 46034231-46034231 on Build GRCh38
rs781322105 Chr.17: 46034316-46034316 on Build GRCh38
rs745815425 Chr.17: 46034154-46034154 on Build GRCh38
rs776853040 Chr.17: 46034300-46034300 on Build GRCh38
rs773932947 Chr.17: 46034187-46034187 on Build GRCh38
rs949553763 Chr.17: 46034152-46034152 on Build GRCh38
rs149756887 Chr.17: 46034199-46034199 on Build GRCh38
rs111514883 Chr.17: 46034286-46034286 on Build GRCh38
rs768459529 Chr.17: 46034186-46034186 on Build GRCh38
rs751853822 Chr.17: 46034134-46034134 on Build GRCh38
rs779914160 Chr.17: 46034327-46034327 on Build GRCh38
rs760065547 Chr.17: 46034256-46034256 on Build GRCh38
rs763362339 Chr.17: 46034280-46034280 on Build GRCh38
rs749039030 Chr.17: 46034184-46034184 on Build GRCh38
rs918091128 Chr.17: 46034146-46034146 on Build GRCh38
rs764568830 Chr.17: 46034283-46034283 on Build GRCh38
rs765884482 Chr.17: 46034257-46034257 on Build GRCh38
rs977852622 Chr.17: 46034133-46034133 on Build GRCh38
rs757306930 Chr.17: 46034135-46034135 on Build GRCh38
rs781138165 Chr.17: 46034153-46034153 on Build GRCh38
rs779806189 Chr.17: 46034167-46034167 on Build GRCh38
rs1054048902 Chr.17: 46034203-46034203 on Build GRCh38
rs751767712 Chr.17: 46034305-46034305 on Build GRCh38
rs375738524 Chr.17: 46034328-46034328 on Build GRCh38
rs757490578 Chr.17: 46034314-46034314 on Build GRCh38
rs756052025 Chr.17: 46034318-46034318 on Build GRCh38
rs138919178 Chr.17: 46034132-46034132 on Build GRCh38
rs769682055 Chr.17: 46034166-46034166 on Build GRCh38

View Full Product Details