Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72346274-72346537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
UniGene
Hs.604479
Gene Chromosome Location
Chr.15: 72343435-72376473 on Build GRCh38
Transcripts
NM_000520.5 NM_001318825.1 NR_134869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6142824 c.1330G>A p.G444S Chr.15: 72346527-72346527 on Build GRCh38
964598 c.1381G>T p.G461* Chr.15: 72346275-72346275 on Build GRCh38
6008077 c.1364G>A p.G455E Chr.15: 72346292-72346292 on Build GRCh38
3816695 c.1339G>C p.E447Q Chr.15: 72346317-72346317 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746814878 Chr.15: 72346349-72346349 on Build GRCh38
rs941638811 Chr.15: 72346482-72346482 on Build GRCh38
rs113941121 Chr.15: 72346297-72346297 on Build GRCh38
rs868661831 Chr.15: 72346515-72346515 on Build GRCh38
rs939203533 Chr.15: 72346319-72346319 on Build GRCh38
rs758066266 Chr.15: 72346317-72346317 on Build GRCh38
rs922317464 Chr.15: 72346376-72346376 on Build GRCh38
rs769826313 Chr.15: 72346377-72346377 on Build GRCh38
rs548496972 Chr.15: 72346409-72346409 on Build GRCh38
rs537494521 Chr.15: 72346505-72346505 on Build GRCh38
rs1029249597 Chr.15: 72346457-72346457 on Build GRCh38
rs778594889 Chr.15: 72346332-72346332 on Build GRCh38
rs754440146 Chr.15: 72346325-72346325 on Build GRCh38
rs1003814544 Chr.15: 72346399-72346399 on Build GRCh38
rs987672900 Chr.15: 72346462-72346462 on Build GRCh38
rs2303448 Chr.15: 72346371-72346371 on Build GRCh38
rs1057519466 Chr.15: 72346307-72346307 on Build GRCh38
rs781126570 Chr.15: 72346359-72346359 on Build GRCh38
rs1016529841 Chr.15: 72346400-72346400 on Build GRCh38
rs866773167 Chr.15: 72346293-72346293 on Build GRCh38
rs1045003375 Chr.15: 72346298-72346298 on Build GRCh38
rs569827588 Chr.15: 72346501-72346501 on Build GRCh38
rs751189932 Chr.15: 72346468-72346468 on Build GRCh38
rs757058366 Chr.15: 72346354-72346354 on Build GRCh38
rs751934469 Chr.15: 72346310-72346310 on Build GRCh38
rs886885437 Chr.15: 72346386-72346386 on Build GRCh38
rs995463313 Chr.15: 72346451-72346451 on Build GRCh38
rs1019027175 Chr.15: 72346471-72346471 on Build GRCh38
rs121907978 Chr.15: 72346296-72346296 on Build GRCh38
rs929725316 Chr.15: 72346391-72346391 on Build GRCh38
rs750903859 Chr.15: 72346320-72346320 on Build GRCh38
rs758099765 Chr.15: 72346344-72346344 on Build GRCh38
rs777608468 Chr.15: 72346347-72346347 on Build GRCh38
rs775571839 Chr.15: 72346480-72346480 on Build GRCh38
rs768970324 Chr.15: 72346338-72346338 on Build GRCh38
rs371435283 Chr.15: 72346511-72346511 on Build GRCh38
rs34085965 Chr.15: 72346318-72346318 on Build GRCh38
rs752346048 Chr.15: 72346337-72346337 on Build GRCh38
rs768838512 Chr.15: 72346495-72346495 on Build GRCh38
rs774507567 Chr.15: 72346499-72346499 on Build GRCh38
rs28940871 Chr.15: 72346305-72346305 on Build GRCh38
rs374588958 Chr.15: 72346485-72346485 on Build GRCh38
rs953668823 Chr.15: 72346458-72346458 on Build GRCh38

View Full Product Details