The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002169.2 | NP_002160.1 | 1 - 1 | 402 | 131 |
GenBank mRNA | BC093755.1 | - | 1 - 1 | 402 | 131 |
BC093757.1 | - | 1 - 1 | 402 | 131 | |
V00541.1 | - | 1 - 1 | 177 | 131 |