The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033257.3 | NP_150282.2 | 3 - 4 | 487 | 73 |
GenBank mRNA | AF228708.1 | - | 3 - 4 | 467 | 73 |
AK001528.1 | - | 3 - 4 | 428 | 73 | |
AK289952.1 | - | 3 - 4 | 405 | 73 | |
AK310899.1 | - | 3 - 4 | 462 | 73 | |
BC000682.2 | - | 3 - 4 | 423 | 73 | |
CR456361.1 | - | 3 - 4 | 392 | 73 |