The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031714.3 | NP_001026884.3 | 21 - 22 | 3833 | 83 |
XM_005268005.4 | XP_005268062.1 | 21 - 22 | 3891 | 83 | |
GenBank mRNA | AL832905.1 | - | 1 - 2 | 311 | 83 |
BC008756.2 | - | 11 - 12 | 1741 | 83 | |
DQ395338.1 | - | 14 - 15 | 2094 | 83 |