The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031714.3 | NP_001026884.3 | 18 - 19 | 2917 | 63 |
NM_022489.3 | NP_071934.3 | 18 - 19 | 2917 | 63 | |
XM_005268004.4 | XP_005268061.1 | 18 - 19 | 3004 | 63 | |
XM_005268005.4 | XP_005268062.1 | 18 - 19 | 2975 | 63 | |
XM_017021595.1 | XP_016877084.1 | 18 - 19 | 2975 | 63 | |
GenBank mRNA | AK025709.1 | - | 9 - 10 | 880 | 63 |
AK225325.1 | - | 9 - 10 | 880 | 63 | |
BC008756.2 | - | 8 - 9 | 825 | 63 | |
BC064828.1 | - | 2 - 3 | 235 | 63 | |
BU538960.1 | - | 4 - 5 | 393 | 63 | |
DQ395338.1 | - | 11 - 12 | 1178 | 63 | |
DQ395339.1 | - | 11 - 12 | 1178 | 63 | |
DQ395340.1 | - | 11 - 12 | 1178 | 63 |