The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032588.3 | NP_115977.2 | 2 - 3 | 478 | 99 |
XM_017002559.1 | XP_016858048.1 | 2 - 3 | 478 | 99 | |
GenBank mRNA | AF353673.1 | - | 2 - 3 | 457 | 99 |
AF361946.1 | - | 2 - 3 | 1282 | 99 | |
AJ276484.1 | - | 2 - 3 | 461 | 99 | |
AK056942.1 | - | 2 - 3 | 474 | 99 | |
BC080529.1 | - | 2 - 3 | 456 | 99 | |
BT007373.1 | - | 2 - 3 | 338 | 99 |