Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD30BL
Entrez Gene ID
554226
Gene Name
ankyrin repeat domain 30B-like
Gene Aliases
ANKRD30BP3, NCRNA00164
Chromosome Location
Chr.2: 132147591 - 132257969 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027019.1 5 - 6 1023 141
GenBank mRNA BC057764.1 - 5 - 6 1026 141

View Full Product Details