Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12192754-12192995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF1B
Entrez Gene ID
7133
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
UniGene
Hs.256278
Gene Chromosome Location
Chr.1: 12166948-12209222 on Build GRCh38
Transcripts
NM_001066.2 XM_011542060.2 XM_011542063.2 XM_017002211.1 XM_017002212.1 XM_017002213.1 XM_017002214.1 XM_017002215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
146406 c.587T>G p.M196R Chr.1: 12192898-12192898 on Build GRCh38
1600661 c.641T>C p.V214A Chr.1: 12192952-12192952 on Build GRCh38
3801492 c.643C>T p.H215Y Chr.1: 12192954-12192954 on Build GRCh38
4504679 c.671C>T p.S224F Chr.1: 12192982-12192982 on Build GRCh38
894799 c.609G>A p.T203T Chr.1: 12192920-12192920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886628953 Chr.1: 12192809-12192809 on Build GRCh38
rs764019731 Chr.1: 12192944-12192944 on Build GRCh38
rs952652900 Chr.1: 12192852-12192852 on Build GRCh38
rs199873050 Chr.1: 12192919-12192919 on Build GRCh38
rs140857317 Chr.1: 12192930-12192930 on Build GRCh38
rs778608285 Chr.1: 12192846-12192847 on Build GRCh38
rs200254914 Chr.1: 12192876-12192876 on Build GRCh38
rs906035722 Chr.1: 12192896-12192896 on Build GRCh38
rs2228494 Chr.1: 12192870-12192870 on Build GRCh38
rs976411216 Chr.1: 12192794-12192794 on Build GRCh38
rs761331191 Chr.1: 12192920-12192920 on Build GRCh38
rs867083487 Chr.1: 12192841-12192841 on Build GRCh38
rs757378017 Chr.1: 12192959-12192959 on Build GRCh38
rs747930689 Chr.1: 12192847-12192849 on Build GRCh38
rs1768642 Chr.1: 12192835-12192835 on Build GRCh38
rs777077445 Chr.1: 12192925-12192925 on Build GRCh38
rs1061622 Chr.1: 12192898-12192898 on Build GRCh38
rs1035218772 Chr.1: 12192913-12192913 on Build GRCh38
rs768027709 Chr.1: 12192899-12192899 on Build GRCh38
rs754426669 Chr.1: 12192821-12192821 on Build GRCh38
rs778099535 Chr.1: 12192851-12192851 on Build GRCh38
rs537233011 Chr.1: 12192884-12192884 on Build GRCh38
rs981800406 Chr.1: 12192904-12192904 on Build GRCh38
rs745706109 Chr.1: 12192970-12192970 on Build GRCh38
rs554329036 Chr.1: 12192822-12192822 on Build GRCh38
rs923246282 Chr.1: 12192807-12192807 on Build GRCh38
rs559822776 Chr.1: 12192789-12192789 on Build GRCh38
rs536867737 Chr.1: 12192869-12192869 on Build GRCh38
rs766601906 Chr.1: 12192817-12192817 on Build GRCh38
rs765420868 Chr.1: 12192823-12192823 on Build GRCh38
rs1047451026 Chr.1: 12192831-12192831 on Build GRCh38
rs964618029 Chr.1: 12192792-12192792 on Build GRCh38
rs759635294 Chr.1: 12192793-12192793 on Build GRCh38
rs549564583 Chr.1: 12192931-12192931 on Build GRCh38
rs745814265 Chr.1: 12192871-12192871 on Build GRCh38
rs373965812 Chr.1: 12192846-12192846 on Build GRCh38
rs780026833 Chr.1: 12192975-12192975 on Build GRCh38
rs771672384 Chr.1: 12192870-12192872 on Build GRCh38
rs759966102 Chr.1: 12192926-12192926 on Build GRCh38
rs751807806 Chr.1: 12192945-12192945 on Build GRCh38
rs781375513 Chr.1: 12192965-12192965 on Build GRCh38
rs757100808 Chr.1: 12192865-12192865 on Build GRCh38
rs775398223 Chr.1: 12192882-12192882 on Build GRCh38
rs747179551 Chr.1: 12192859-12192859 on Build GRCh38
rs201545665 Chr.1: 12192924-12192924 on Build GRCh38
rs758561163 Chr.1: 12192933-12192933 on Build GRCh38

View Full Product Details