The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030981.2 | NP_112243.1 | 4 - 5 | 376 | 91 |
XM_017018378.1 | XP_016873867.1 | 4 - 5 | 435 | 91 | |
GenBank mRNA | AF092437.1 | - | 4 - 5 | 294 | 91 |
AJ245875.1 | - | 3 - 4 | 281 | 93 | |
AK292086.1 | - | 4 - 5 | 344 | 91 | |
AK315333.1 | - | 4 - 5 | 337 | 91 | |
AL136635.1 | - | 4 - 5 | 328 | 91 | |
BC028331.1 | - | 4 - 5 | 327 | 91 | |
BC071169.1 | - | 4 - 5 | 315 | 91 | |
CR533462.1 | - | 3 - 4 | 281 | 91 | |
U66621.1 | - | 1 - 2 | 84 | 91 |