Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YLPM1
Entrez Gene ID
56252
Gene Name
YLP motif containing 1
Gene Aliases
C14orf170, PPP1R169, ZAP113, ZAP3
Chromosome Location
Chr.14: 74763322 - 74837310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019589.2 NP_062535.2 15 - 16 6073 102
XM_005267860.3 XP_005267917.1 15 - 16 6117 102
XM_011536966.2 XP_011535268.1 16 - 17 5232 102
XM_011536967.1 XP_011535269.1 14 - 15 3999 102
GenBank mRNA AK090435.1 - 13 - 14 4829 102
AK297582.1 - 11 - 12 1644 102
AL832365.1 - 12 - 13 4608 102
BC007792.1 - 6 - 7 670 102
BC023570.2 - 2 - 3 160 102
L40403.1 - 9 - 10 1177 102

View Full Product Details