The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_111966.1 | 2 - 3 | 683 | 119 | |
NR_111967.1 | 2 - 3 | 663 | 119 | ||
GenBank mRNA | AK001276.1 | - | 2 - 3 | 401 | 119 |
BC024245.2 | - | 2 - 3 | 476 | 119 | |
BC090958.1 | - | 2 - 3 | 483 | 119 | |
BI823697.1 | - | 2 - 3 | 483 | 119 | |
DT216192.1 | - | 2 - 3 | 359 | 119 |