The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005246263.2 | XP_005246320.1 | 26 - 27 | 3693 | 82 |
XM_011510531.1 | XP_011508833.1 | 27 - 28 | 3714 | 82 | |
GenBank mRNA | AK131316.1 | - | 12 - 13 | 1824 | 82 |