The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267782.1 | NP_001254711.1 | 3 - 4 | 554 | 95 |
NM_001267783.1 | NP_001254712.1 | 3 - 4 | 554 | 95 | |
NM_001300731.1 | NP_001287660.1 | 3 - 4 | 554 | 95 | |
NM_017749.3 | NP_060219.2 | 3 - 4 | 552 | 95 | |
XM_005253009.3 | XP_005253066.1 | 3 - 4 | 550 | 95 | |
XM_005253011.3 | XP_005253068.1 | 3 - 4 | 550 | 95 | |
XM_005253014.3 | XP_005253071.1 | 3 - 4 | 550 | 95 | |
XM_006718259.2 | XP_006718322.1 | 3 - 4 | 431 | 95 | |
XM_006718260.2 | XP_006718323.1 | 3 - 4 | 550 | 95 | |
XM_017018007.1 | XP_016873496.1 | 3 - 4 | 550 | 95 | |
XM_017018008.1 | XP_016873497.1 | 3 - 4 | 550 | 95 | |
GenBank mRNA | AB051523.1 | - | 3 - 4 | 506 | 95 |
AK302808.1 | - | 3 - 4 | 612 | 95 | |
AL834190.1 | - | 3 - 4 | 433 | 95 | |
BC045609.1 | - | 3 - 4 | 552 | 95 | |
DQ870924.1 | - | 2 - 3 | 192 | 95 |