The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297713.1 | NP_001284642.1 | 7 - 8 | 2689 | 76 |
NM_015441.2 | NP_056256.1 | 7 - 8 | 2686 | 76 | |
XM_005245075.2 | XP_005245132.1 | 7 - 8 | 2692 | 76 | |
XM_011509398.2 | XP_011507700.1 | 4 - 5 | 2847 | 76 | |
GenBank mRNA | AB119054.1 | - | 7 - 8 | 1651 | 76 |
AK301499.1 | - | 7 - 8 | 1864 | 76 | |
AK316154.1 | - | 7 - 8 | 2018 | 76 | |
AL050137.1 | - | 2 - 3 | 699 | 76 | |
BC047472.1 | - | 1 - 2 | 429 | 76 | |
BC067274.1 | - | 7 - 8 | 2075 | 76 | |
BX648975.1 | - | 7 - 8 | 1862 | 76 |