Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AMIGO2
Entrez Gene ID
347902
Gene Name
adhesion molecule with Ig like domain 2
Gene Aliases
ALI1, AMIGO-2, DEGA
Chromosome Location
Chr.12: 47075707 - 47080826 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181847.4 NP_862830.1 1 - 2 490 115
GenBank mRNA AK091761.1 - 1 - 2 313 115
AY454159.1 - 1 - 2 405 115
BC047595.1 - 1 - 2 405 115

View Full Product Details