The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110514.1 | NP_001103984.1 | 16 - 17 | 1996 | 147 |
XM_006723600.1 | XP_006723663.1 | 16 - 17 | 1791 | 147 | |
XM_017027983.1 | XP_016883472.1 | 17 - 18 | 1888 | 147 | |
XM_017027984.1 | XP_016883473.1 | 17 - 18 | 1994 | 147 | |
XM_017027985.1 | XP_016883474.1 | 10 - 11 | 1150 | 147 | |
GenBank mRNA | BC019106.2 | - | 2 - 3 | 487 | 147 |
BC037555.1 | - | 8 - 9 | 1096 | 147 | |
BC054347.1 | - | 1 - 2 | 650 | 147 |