The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016067.3 | NP_057151.1 | 2 - 3 | 294 | 141 |
GenBank mRNA | AF151892.1 | - | 2 - 3 | 207 | 141 |
AK026620.1 | - | 2 - 3 | 261 | 141 | |
BC005186.1 | - | 2 - 3 | 185 | 141 |