The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197259.1 | NP_001184188.1 | 12 - 13 | 1381 | 149 |
NM_012381.3 | NP_036513.2 | 13 - 14 | 1479 | 149 | |
NM_181837.2 | NP_862820.1 | 13 - 14 | 1479 | 149 | |
XM_005248704.1 | XP_005248761.1 | 13 - 14 | 1425 | 149 | |
XM_005248705.1 | XP_005248762.1 | 14 - 15 | 1483 | 149 | |
XM_005248706.1 | XP_005248763.1 | 14 - 15 | 1482 | 149 | |
XM_011535651.1 | XP_011533953.1 | 12 - 13 | 1311 | 149 | |
XM_017010632.1 | XP_016866121.1 | 13 - 14 | 1425 | 149 | |
XM_017010633.1 | XP_016866122.1 | 13 - 14 | 1425 | 149 | |
XM_017010634.1 | XP_016866123.1 | 13 - 14 | 1425 | 149 | |
XM_017010635.1 | XP_016866124.1 | 12 - 13 | 1311 | 149 | |
XM_017010636.1 | XP_016866125.1 | 14 - 15 | 1483 | 149 | |
XM_017010637.1 | XP_016866126.1 | 14 - 15 | 1482 | 149 | |
XM_017010638.1 | XP_016866127.1 | 14 - 15 | 1483 | 149 | |
XM_017010639.1 | XP_016866128.1 | 14 - 15 | 1482 | 149 | |
XM_017010640.1 | XP_016866129.1 | 13 - 14 | 1368 | 149 | |
GenBank mRNA | AF093535.1 | - | 13 - 14 | 1403 | 149 |
AF125507.1 | - | 13 - 14 | 1412 | 149 | |
AF135044.1 | - | 13 - 14 | 1377 | 149 | |
AK303056.1 | - | 13 - 14 | 1405 | 149 | |
AK303182.1 | - | 12 - 13 | 1200 | 149 | |
AK303195.1 | - | 12 - 13 | 1305 | 149 | |
AK315911.1 | - | 13 - 14 | 1366 | 149 | |
AK316298.1 | - | 13 - 14 | 1410 | 149 | |
AL080116.1 | - | 13 - 14 | 1418 | 149 | |
BC035494.2 | - | 13 - 14 | 1394 | 149 | |
BC047689.1 | - | 13 - 14 | 1395 | 149 | |
CR456814.1 | - | 8 - 9 | 912 | 149 | |
U50950.1 | - | 9 - 10 | 1045 | 149 |