The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014508.2 | NP_055323.2 | 1 - 2 | 118 | 110 |
GenBank mRNA | AB208979.1 | - | 1 - 2 | 83 | 110 |
AF165520.1 | - | 1 - 2 | 122 | 110 | |
AK094570.1 | - | - | 1432 | 110 | |
AK313272.1 | - | 1 - 2 | 122 | 110 | |
BC011739.2 | - | 1 - 2 | 118 | 110 | |
BC021080.1 | - | 1 - 2 | 105 | 110 | |
CR456394.1 | - | 1 - 2 | 96 | 110 |