Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMED10
Entrez Gene ID
10972
Gene Name
transmembrane p24 trafficking protein 10
Gene Aliases
P24(DELTA), S31I125, S31III125, TMP21, Tmp-21-I, p23, p24d1
Chromosome Location
Chr.14: 75131468 - 75176646 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006827.5 NP_006818.3 1 - 1 63 94
GenBank mRNA AK296800.1 - 1 - 1 32 94
AK312384.1 - 1 - 1 32 94
BC001496.2 - 1 - 1 48 94
BC001825.2 - 1 - 1 48 94
BX504393.1 - 1 - 1 64 94

View Full Product Details