The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267817.1 | NP_001254746.1 | 3 - 3 | 669 | 177 |
NM_001267818.1 | NP_001254747.1 | 5 - 5 | 941 | 177 | |
NM_021227.3 | NP_067050.1 | 4 - 4 | 867 | 177 | |
GenBank mRNA | AF068297.1 | - | 4 - 4 | 830 | 177 |
AF343342.1 | - | 4 - 4 | 796 | 177 | |
BC016321.1 | - | 4 - 4 | 855 | 177 | |
BC054857.1 | - | 4 - 4 | 857 | 177 | |
BM994257.1 | - | 1 - 1 | 177 | ||
CD369095.1 | - | 1 - 1 | 177 |