Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
BRD7
Entrez Gene ID
29117
Gene Name
bromodomain containing 7
Gene Aliases
BP75, CELTIX1, NAG4
Chromosome Location
Chr.16: 50317563 - 50368969 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173984.2 NP_001167455.1 15 - 15 1838 123
NM_013263.4 NP_037395.2 15 - 15 1835 123
XM_011523046.2 XP_011521348.1 15 - 15 1924 123
XM_011523047.2 XP_011521349.1 15 - 15 1921 123
XM_011523048.2 XP_011521350.1 14 - 14 1816 123
XM_011523049.2 XP_011521351.1 14 - 14 1813 123
XM_011523050.2 XP_011521352.1 11 - 11 1944 123
XM_017023179.1 XP_016878668.1 14 - 14 1630 123
XM_017023180.1 XP_016878669.1 14 - 14 1627 123
XM_017023181.1 XP_016878670.1 11 - 11 1958 123
GenBank mRNA AF152604.1 - 15 - 15 1728 123
AF213969.1 - 15 - 15 1683 123
AJ271881.1 - 15 - 15 1793 123
AK027308.1 - 8 - 8 806 123
AK074613.1 - 9 - 9 972 123
BC001611.1 - 11 - 11 1099 123
BC050728.1 - 16 - 16 1742 123
BC094706.1 - 16 - 16 1712 123

View Full Product Details