Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLGA8M
Entrez Gene ID
653720
Gene Name
golgin A8 family member M
Gene Aliases
-
Chromosome Location
Chr.15: 28698695 - 28718512 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282468.1 NP_001269397.1 8 - 9 690 65
XM_005268284.4 XP_005268341.1 8 - 9 1489 65
XM_005268285.4 XP_005268342.1 8 - 9 1489 65
XM_006720654.3 XP_006720717.1 8 - 9 633 65
XM_011521951.2 XP_011520253.1 9 - 10 1058 65
XM_011521952.2 XP_011520254.1 8 - 9 1489 65
XM_011521953.2 XP_011520255.1 8 - 9 1489 65
XM_011521954.2 XP_011520256.1 8 - 9 623 65

Gene Symbol
GOLGA8IP
Entrez Gene ID
283796
Gene Name
golgin A8 family member I, pseudogene
Gene Aliases
GOLGA8I, GOLGA9P
Chromosome Location
Chr.15: 22610353 - 22617854 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_024074.1 7 - 8 891 65
GenBank mRNA AK093104.1 - 7 - 8 891 65

View Full Product Details