Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C17orf58
Entrez Gene ID
284018
Gene Name
chromosome 17 open reading frame 58
Gene Aliases
-
Chromosome Location
Chr.17: 67991099 - 67993649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181656.4 NP_858042.2 2 - 3 563 113
GenBank mRNA AK026583.1 - 2 - 3 501 113
BC021702.1 - 1 - 2 182 113
BX364842.2 - 2 - 3 562 113

View Full Product Details