The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012186.2 | NP_036318.1 | 1 - 1 | 1839 | 131 |
GenBank mRNA | AF275722.1 | - | 1 - 1 | 1850 | 131 |