The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031439.3 | NP_113627.1 | 2 - 2 | 1836 | 114 |
GenBank mRNA | AK055556.1 | - | 2 - 2 | 1774 | 114 |
BC004299.1 | - | 2 - 2 | 1763 | 114 | |
BC071947.1 | - | 2 - 2 | 1762 | 114 | |
BU187815.1 | - | 1 - 1 | 155 | 114 |