The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031439.3 | NP_113627.1 | 2 - 2 | 2706 | 168 |
GenBank mRNA | BG740632.1 | - | 1 - 1 | 273 | 168 |