The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000994.3 | NP_000985.1 | 4 - 4 | 498 | 146 |
NM_001007073.1 | NP_001007074.1 | 4 - 4 | 597 | 146 | |
NM_001007074.1 | NP_001007075.1 | 3 - 3 | 617 | 146 | |
GenBank mRNA | BQ068496.1 | - | - | 535 | 146 |
X03342.1 | - | 4 - 4 | 438 | 146 |