The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013240.5 | NP_037372.4 | 6 - 6 | 727 | 138 |
NM_182749.4 | NP_877426.4 | 5 - 5 | 643 | 138 | |
GenBank mRNA | AF139682.1 | - | 6 - 6 | 728 | 138 |
AW136986.1 | - | 1 - 1 | 138 | ||
BC011554.1 | - | 5 - 5 | 624 | 138 |