The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003754.2 | NP_003745.1 | 1 - 2 | 399 | 82 |
GenBank mRNA | AK093511.1 | - | 3 - 4 | 991 | 82 |
AK095574.1 | - | 2 - 3 | 361 | 82 | |
AK289637.1 | - | 1 - 2 | 382 | 82 | |
AK291354.1 | - | 1 - 2 | 379 | 82 | |
AK297621.1 | - | 2 - 3 | 232 | 82 | |
BC000490.2 | - | 1 - 1 | 372 | 82 | |
BC073753.1 | - | 1 - 2 | 367 | 82 | |
BT006894.1 | - | 1 - 2 | 366 | 82 | |
CB141463.1 | - | 1 - 2 | 399 | 82 | |
CD050033.1 | - | 1 - 2 | 399 | 82 | |
CR456959.1 | - | 1 - 2 | 366 | 82 | |
U94855.1 | - | 1 - 2 | 372 | 82 |