Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
DEPDC1
Entrez Gene ID
55635
Gene Name
DEP domain containing 1
Gene Aliases
DEP.8, DEPDC1-V2, DEPDC1A, SDP35
Chromosome Location
Chr.1: 68474152 - 68497221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114120.2 NP_001107592.1 2 - 2 294 102
NM_017779.5 NP_060249.2 2 - 2 294 102
GenBank mRNA AB281187.1 - 2 - 2 150 102
AB281274.1 - 2 - 2 150 102
AJ278112.1 - 2 - 2 150 102
AK000361.1 - 2 - 2 155 102
AK225735.1 - 2 - 2 140 102
BC003511.1 - 2 - 2 143 102
BC065304.1 - 2 - 2 97 102
DB147703.1 - 2 - 2 294 102

View Full Product Details