The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039503.2 | NP_001034592.1 | 4 - 5 | 510 | 62 |
XM_011545819.1 | XP_011544121.1 | 4 - 5 | 1736 | 62 | |
XM_011545820.1 | XP_011544122.1 | 4 - 5 | 1736 | 62 | |
GenBank mRNA | AJ627035.1 | - | 4 - 5 | 510 | 62 |