Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAMR1
Entrez Gene ID
25891
Gene Name
peptidase domain containing associated with muscle regeneration 1
Gene Aliases
DKFZP586H2123, FP938, RAMP
Chromosome Location
Chr.11: 35431827 - 35530300 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001991.2 NP_001001991.1 3 - 4 832 75
NM_001282675.1 NP_001269604.1 5 - 6 822 75
NM_015430.3 NP_056245.2 3 - 4 832 75
GenBank mRNA AF370388.1 - 2 - 3 320 75
AK027841.1 - 3 - 4 432 75
AK297092.1 - 5 - 6 823 75
AL832391.1 - 3 - 4 398 75
AY358346.1 - 3 - 4 411 75
BC089434.1 - 3 - 4 424 75
BX640676.1 - 3 - 4 832 75

View Full Product Details