The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001991.2 | NP_001001991.1 | 4 - 5 | 945 | 117 |
NM_001282675.1 | NP_001269604.1 | 6 - 7 | 935 | 117 | |
NM_015430.3 | NP_056245.2 | 4 - 5 | 945 | 117 | |
GenBank mRNA | AF370388.1 | - | 3 - 4 | 433 | 117 |
AK027841.1 | - | 4 - 5 | 545 | 117 | |
AK297092.1 | - | 6 - 7 | 936 | 117 | |
AL832391.1 | - | 4 - 5 | 511 | 117 | |
AY358346.1 | - | 4 - 5 | 117 | ||
BC089434.1 | - | 4 - 5 | 537 | 117 | |
BX640676.1 | - | 4 - 5 | 945 | 117 |