The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005254042.1 | XP_005254099.1 | 3 - 4 | 553 | 110 |
GenBank mRNA | AY170470.2 | - | 4 - 5 | 417 | 110 |
AY170471.1 | - | 3 - 4 | 295 | 110 | |
AY223901.1 | - | 3 - 4 | 553 | 110 |